Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RB Antibody (938951) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Specifications
Antigen | IL-17RB |
---|---|
Clone | 938951 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry, CyTOF-ready |
Applications | Flow Cytometry, CyTOF |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB12074SP
![]() |
Novus Biologicals
MAB12074SP |
25 μg | N/A | N/A | N/A | ||||
MAB12074
![]() |
Novus Biologicals
MAB12074 |
100 μg | N/A | N/A | N/A | ||||
Description
IL-17RB Monoclonal specifically detects IL-17RB in Human samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
IL-17RB | |
1.0 mg/mL | |
Flow Cytometry, CyTOF | |
Unconjugated | |
Mouse | |
Cytokine Research | |
PBS with No Preservative | |
55540 | |
Partial recombinant human IL-17RB protein (aa18-289) | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
938951 | |
Flow Cytometry, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CRL4IL-17 receptor homolog 1, cytokine receptor CRL4, Cytokine receptor-like 4, EVI27IL-17B receptor, IL-17 receptor B, IL17BRinterleukin 17B receptor, IL-17RB, IL17Rh1, IL-17Rh1, IL17RH1MGC5245, interleukin 17 receptor B, interleukin 17 receptor homolog 1, interleukin-17 receptor B, Interleukin-17B receptor | |
IL17RB | |
IgG1 | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title