Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RB Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984849
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4849 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4849 Supplier Bioss Supplier No. BS2610R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recently, a number of cytokines belonging to the interleukin (IL)-17 family have been identified. These are termed as IL-17B, IL-17C (1) and IL-17E. IL-17 is a potent proinflammatory cytokine that plays roles in a number of diseases including rheumatoid arthritis, multiple sclerosis, and promotion of tumor growth. IL-17B, C, and E like IL-17 are able to induce proinflammatory responses. However, they do not bind to the IL-17 receptor suggesting that additional IL-17R related receptor might exist. Receptor for IL-17B and IL-17E has been independently isolated and has been designated as EV127 (in mouse) and IL-17Rh1 (in human), respectively. IL-17E induces activation of NF-kB pathway and like IL-17 also induces production of IL-8. IL-17Rh1 mRNA is highly expressed in Kidney and liver and at moderate level in small intestine, colon, and brain.

Specifications

Antigen IL-17RB
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IL17RB
Gene Accession No. Q9NRM6
Gene Alias CRL4; cytokine receptor CRL4; Cytokine receptor-like 4; ecotropic viral integration site 27; Evi27; IL-17 receptor B; IL-17 receptor homolog 1; IL-17B; IL-17B receptor; Il17br; IL-17ER; IL17RB; IL-17RB; IL17RH1; IL-17Rh1; IL-20; interleukin 17 receptor B; interleukin 17 receptor homolog 1; interleukin 17B receptor; Interleukin-17 receptor B; Interleukin-17B receptor; MGC138900; MGC138901; MGC5245; UNQ2501/PRO19612; ZCYTO7
Gene Symbols IL17RB
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IL-17RB.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55540
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.