Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RC Antibody (209), DyLight 755 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289976IR
Description
IL-17RC Monoclonal antibody specifically detects IL-17RC in Human samples. It is validated for ELISASpecifications
IL-17RC | |
Monoclonal | |
DyLight 755 | |
FLJ95963, FLJ96005, IL-17 receptor C, IL17F receptor, IL-17RC, IL17Rhom, IL17RL, IL-17RL, IL17-RL, interleukin 17 receptor C, interleukin-17 receptor C, Interleukin-17 receptor homolog, Interleukin-17 receptor-like protein, MGC10763, zcytoR14 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-17RC (NP_116121.2; Met1-Ala454). | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
209 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
84818 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction