Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RD/SEF Antibody (400210), mFluor Violet 500 SE, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2276MFV500
Description
IL-17RD/SEF Monoclonal antibody specifically detects IL-17RD/SEF in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOFSpecifications
| IL-17RD/SEF | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| 400210 | |
| Flow Cytometry, Immunohistochemistry, CyTOF-ready | |
| FLJ35755, IL-17RDIL-17 receptor D, IL17Rhom, IL17RLMDKFZp434N1928, interleukin 17 receptor D, interleukin-17 receptor D, Interleukin-17 receptor-like protein, MGC133309, Sef homolog, SEFhSef | |
| Mouse myeloma cell line NS0-derived recombinant mouse IL-17 RD/SEF, Gly28-Arg299, Accession # Q8JZL1 | |
| RUO | |
| 54756 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction