Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RE Antibody (46N8G7), Alexa Fluor™ 532, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP227366AF532

Catalog No. NP227366A32


Only null left
Add to Cart

Description

Description

IL-17RE Monoclonal specifically detects IL-17RE in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-17RE
Monoclonal
Alexa Fluor 532
50 mM sodium borate with 0.05% sodium azide
IL17RE
amino acids (97-199) MVHLLVQkSkkSSTFkFYRRHkMPAPAQRkLLPRRHLSEkSHHISIPSPDISHkGLRSkR∼TQPSDPETWESLPRLDSQRHGGPEFSFDLLPEARAIRVTISSGPE
Primary
Human
IgG1 κ
Western Blot
46N8G7
Western Blot
IL-17 receptor E, interleukin 17 receptor E
Mouse
0.1 mL
132014
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.