Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RE Antibody (46N8G7), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP227366FR
Description
IL-17RE Monoclonal specifically detects IL-17RE in Human samples. It is validated for Western Blot.Specifications
IL-17RE | |
Monoclonal | |
DyLight 680 | |
Q8NFR9 | |
IL17RE | |
amino acids (97-199) MVHLLVQkSkkSSTFkFYRRHkMPAPAQRkLLPRRHLSEkSHHISIPSPDISHkGLRSkR∼TQPSDPETWESLPRLDSQRHGGPEFSFDLLPEARAIRVTISSGPE | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
46N8G7 | |
Western Blot | |
IL-17 receptor E, interleukin 17 receptor E | |
Mouse | |
Protein G purified | |
RUO | |
132014 | |
Store at 4C in the dark. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction