Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-17RE Antibody (46N8G7), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$181.90 - $366.00

Specifications

Antigen IL-17RE
Clone 46N8G7
Concentration 0.5 mg/mL
Dilution Western Blot 5-10ug/ml∼
Applications Western Blot
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP227365SS
SDP
View Documents
Novus Biologicals
NBP227365SS
0.025 mg
Each for $181.90
Only null left
Add to Cart
 
NBP227365
SDP
View Documents
Novus Biologicals
NBP227365
0.1 mg
Each for $366.00
Only null left
Add to Cart
 
Description

Description

IL-17RE Monoclonal specifically detects IL-17RE in Human samples. It is validated for Western Blot.
Specifications

Specifications

IL-17RE
0.5 mg/mL
Western Blot
Unconjugated
Mouse
Human
IL-17 receptor E, interleukin 17 receptor E
IL17RE
IgG1 κ
Protein G purified
46N8G7
Western Blot 5-10ug/ml∼
Monoclonal
Purified
RUO
Q8NFR9
132014
amino acids (97-199) MVHLLVQkSkkSSTFkFYRRHkMPAPAQRkLLPRRHLSEkSHHISIPSPDISHkGLRSkR∼TQPSDPETWESLPRLDSQRHGGPEFSFDLLPEARAIRVTISSGPE
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.