Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-17RE Antibody (944904), DyLight 755, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB7997Z
Description
IL-17RE Monoclonal antibody specifically detects IL-17RE in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
IL-17RE | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
132014 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, CyTOF | |
944904 | |
Flow Cytometry, CyTOF-ready | |
IL-17 receptor E, interleukin 17 receptor E | |
Mouse myeloma cell line NSO-derived recombinant mouse IL-17 RE, Met1-His414, Accession # Q8BH06 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction