Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18 BPa/IL18BP Antibody (224), Alexa Fluor™ 350 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289445AF350
Description
IL-18 BPa/IL18BP Monoclonal antibody specifically detects IL-18 BPa/IL18BP in Human samples. It is validated for ELISASpecifications
IL-18 BPa/IL18BP | |
Monoclonal | |
Alexa Fluor 350 | |
IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-18 BPa/IL18BP (NP_001034748.1; Met 1-Gly 192). | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity | |
10068 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
224 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction