Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18 BPa/IL18BP Antibody (224), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289445F
Description
IL-18 BPa/IL18BP Monoclonal antibody specifically detects IL-18 BPa/IL18BP in Human samples. It is validated for ELISASpecifications
| IL-18 BPa/IL18BP | |
| Monoclonal | |
| FITC | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 224 | |
| ELISA | |
| IL-18BP, IL18BPa, interleukin 18 binding protein, interleukin-18-binding protein, MC51L-53L-54L homolog gene product, tadekinig-alfa | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-18 BPa/IL18BP (NP_001034748.1; Met 1-Gly 192). | |
| 0.1 mL | |
| Cytokine Research, Immunology, Innate Immunity | |
| 10068 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction