Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse IL-18 BPd Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF122SP
This item is not returnable.
View return policy
Description
IL-18 BPd Polyclonal specifically detects IL-18 BPd in Mouse samples. It is validated for Western Blot, ELISA, Neutralization.Specifications
IL-18 BPd | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Il18bp | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-18 BPd Thr27-Ala191 Accession # AAD17194 | |
25 μg | |
Primary | |
Detects mouse IL-18 BPd in direct ELISAs and Western blots. | |
Mouse | |
IgG |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 0.1 ug/mL, Neutralization 10-30 ug/mL | |
AAD17194 | |
Goat | |
Affinity Purified | |
RUO | |
16068 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction