Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18/IL-1F4 Antibody (125-2H), DyLight 650, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281040C
Description
IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
IL-18/IL-1F4 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate | |
IL18 | |
Fusion of splenocytes from mice immunized with human IL-18/IL-1F4, with SP2/O mouse myeloma cells. | |
0.1 mL | |
Primary | |
Recognizes the active form of human IL-18/IL-1F4. | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
125-2H | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
Rabbit | |
Protein A purified | |
RUO | |
3606 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction