Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18/IL-1F4 Antibody (167), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290394AF647
Description
IL-18/IL-1F4 Monoclonal specifically detects IL-18/IL-1F4 in Mouse samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
IL-18/IL-1F4 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
3606 | |
Mouse | |
Purified |
ELISA | |
167 | |
ELISA, Sandwich ELISA Detection | |
Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-18/IL-1F4 (P70380; Asn36-Ser192). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human IL1F7 Human IL1F9 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction