Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18/IL-1F4 Antibody (169), Janelia Fluor™ 549 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290395JF549
Description
IL-18/IL-1F4 Monoclonal antibody specifically detects IL-18/IL-1F4 in Mouse samples. It is validated for ELISA, ELISASpecifications
IL-18/IL-1F4 | |
Monoclonal | |
Janelia Fluor 549 | |
Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-18/IL-1F4 (P70380; Asn36-Ser192). | |
0.1 mL | |
Angiogenesis, Apoptosis, Cancer, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Endocrinology, Immunology, Innate Immunity | |
3606 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
169 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction