Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18/IL-1F4 Antibody (74801), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB646P
Description
IL-18/IL-1F4 Monoclonal antibody specifically detects IL-18/IL-1F4 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF, KnockDownSpecifications
IL-18/IL-1F4 | |
Monoclonal | |
PE | |
PBS | |
Mouse | |
Protein A or G purified from ascites | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Flow Cytometry, CyTOF, KnockDown | |
74801 | |
Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready, Knockout Validated | |
Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
E. coli-derived recombinant human IL-18/IL-1F4, Accession # Q14116 | |
0.5 mL | |
Angiogenesis, Apoptosis, Cancer, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Endocrinology, Immunology, Innate Immunity | |
3606 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction