Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18/IL-1F4 Antibody (925008), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB2548C
Description
IL-18/IL-1F4 Monoclonal antibody specifically detects IL-18/IL-1F4 in Human samples. It is validated for ELISA, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
IL-18/IL-1F4 | |
Monoclonal | |
PerCP | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
925008 | |
ELISA, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
Iboctadekin, IFN-gamma-inducing factor, IGIFIL-1 gamma, IL-18MGC12320, IL1F4iboctadekin, IL-1g, Interferon gamma-inducing factor, interleukin 18 (interferon-gamma-inducing factor), Interleukin-1 gamma, interleukin-18 | |
E. coli-derived recombinant Rhesus monkey IL-18/IL-1F4, Tyr37-Asp193, Accession # Q14116 | |
0.1 mL | |
Angiogenesis, Apoptosis, Cancer, Cardiovascular Biology, Cell Biology, Cytokines and Growth Factors, Endocrinology, Immunology, Innate Immunity | |
3606 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction