Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18 Monoclonal Antibody (C10), Invitrogen™
Mouse Monoclonal Antibody
$184.40 - $522.50
Specifications
Antigen | IL-18 |
---|---|
Clone | C10 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA543741
|
Invitrogen™
MA543741 |
20 μL |
Each for $184.40
|
|
|||||
PIMA543740
|
Invitrogen™
MA543740 |
100 μL |
Each for $522.50
|
|
|||||
Description
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.Specifications
IL-18 | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320 | |
IL18 | |
IgG1 κ | |
Protein A/G | |
Antibody |
C10 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human, Rat, Pig | |
O19073, P97636, Q14116 | |
29197, 3606, 397057 | |
Arg94-Asp193 (Accession Q14116), with N-terminal His Tag. | |
Primary | |
-20°C | |
IL18 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title