Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-18 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547344
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547344 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547344 Supplier Invitrogen™ Supplier No. PA547344
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody detects rhesus macaque IL-18/IL-1F4 in direct ELISAs and Western blots. In Western blots, approximately 10% cross-reactivity with recombinant mouse IL-18, recombinant rat IL-18, and recombinant porcine IL-18 is observed. Reconstitute at 0.2 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 μg of the antibody by the LAL method.

The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-18
Applications ELISA, Neutralization, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene IL18
Gene Accession No. Q14116
Gene Alias cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320
Gene Symbols IL18
Host Species Goat
Immunogen E. coli-derived recombinant rhesus macaque IL-18/IL-1F4 Tyr37-Asp193.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3606, 574151
Target Species Human, Rhesus Macaque
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.