Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18 R beta/IL-1 R7/ACPL Antibody (290), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289317AF750
Description
IL-18 R beta/IL-1 R7/ACPL Monoclonal specifically detects IL-18 R beta/IL-1 R7/ACPL in Human samples. It is validated for Western Blot, ELISA.Specifications
IL-18 R beta/IL-1 R7/ACPL | |
Monoclonal | |
Alexa Fluor 750 | |
Accessory protein-like, ACPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-18 receptor accessory protein, IL-18 receptor beta, IL-18RAcP, IL18RB, IL-18Rbeta, IL-18R-beta, IL-1R accessory protein-like, IL1R7, IL-1R7, IL-1R-7, IL-1RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-18 R beta/IL-1 R7/ACPL (NP_003844.1; Met1-Gly357). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, ELISA | |
290 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
8807 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction