Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18 R beta/IL-1 R7/ACPL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 1 publication
$181.90 - $487.50
Specifications
| Antigen | IL-18 R beta/IL-1 R7/ACPL |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 1-3 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056433T
![]() |
Novus Biologicals
NB10056433SS |
0.025 mg |
Each for $181.90
|
|
|||||
NB10056433
![]() |
Novus Biologicals
NB10056433 |
0.1 mg |
Each for $487.50
|
|
|||||
Description
IL-18 R beta/IL-1 R7/ACPL Polyclonal specifically detects IL-18 R beta/IL-1 R7/ACPL in Human, Mouse samples. It is validated for Western Blot.Specifications
| IL-18 R beta/IL-1 R7/ACPL | |
| Western Blot 1-3 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS containing 0.05% BSA with 0.05% Sodium Azide | |
| Accessory protein-like, ACPL, CD218 antigen-like family member B, CD218b, CD218b antigen, CDw218b, IL-18 receptor accessory protein, IL-18 receptor beta, IL-18RAcP, IL18RB, IL-18Rbeta, IL-18R-beta, IL-1R accessory protein-like, IL1R7, IL-1R7, IL-1R-7, IL-1RAcPL, interleukin 18 receptor accessory protein, Interleukin-1 receptor 7, interleukin-18 receptor accessory protein, Interleukin-18 receptor accessory protein-like, Interleukin-18 receptor beta, MGC120589, MGC120590 | |
| IL18RAP | |
| IgG | |
| Protein G purified |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O95256 | |
| 8807 | |
| A synthetic peptide corresponding to a region within amino acids 50-150 of human IL-18 Receptor accessory protein/IL-18 RA/AcPL was used immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title