Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL-18 Recombinant Mouse Monoclonal Antibody (125-2H)
Mouse Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA547794
Description
Specificity: Recognizes the active form of human IL-18.
The protein encoded by this gene is a proinflammatory cytokine. This cytokine can induce the IFN-gamma production of T cells. The combination of this cytokine and IL12 has been shown to inhibit IL4 dependent IgE and IgG1 production, and enhance IgG2a production of B cells. IL-18 binding protein can specifically interact with this cytokine, and thus negatively regulate its biological activity.
Specifications
IL-18 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.02% ProClin 300 | |
Q14116 | |
IL18 | |
Fusion of splenocytes from mice immunized with human IL-18, with SP2/O mouse myeloma cells. | |
200 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Immunoprecipitation, Neutralization, Immunocytochemistry | |
125-2H | |
Unconjugated | |
IL18 | |
cytokine; iboctadekin; IFN-gamma-inducing factor; IGIF; il 18; IL-1 gamma; IL18; IL-18; IL1F4; IL-1g; ILN; interferon gamma inducing factor; interferon gamma-inducing factor; interferon-gamma-inducing factor; Interleukin; interleukin 18; interleukin 18 (interferon-gamma-inducing factor); interleukin-1 gamma; Interleukin18; Interleukin-18; interleukin-18 proprotein; interleukin-18-like protein; MGC12320 | |
Mouse | |
Protein A | |
RUO | |
3606 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction