Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IL-18BP Rabbit anti-Human, Mouse, Rat, Polyclonal, ProteinTech
SDP

Catalog No. 502238326 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-8326 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-8326 Supplier Proteintech Group Inc Supplier No. 141531AP150UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Aliquoting is unnecessary for -20°C storage.

IL-18BP Polyclonal antibody specifically detects IL-18BP in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen IL-18BP
Applications Western Blot
Classification Polyclonal
Concentration 300 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene IL18BP
Gene Accession No. O95998, Q9Z0M9
Gene Alias Igifbp; Il18bp; IL-18BP; IL18BPa; interferon gamma inducing factor binding protein; interferon gamma-inducing factor-binding protein; interleukin 18 binding protein; interleukin-18 binding protein; interleukin-18-binding protein; MC51L-53L-54L homolog gene product; MC54L; Tadekinig-alfa
Gene Symbols IL18BP
Host Species Rabbit
Immunogen IL-18BP Fusion Protein Ag5335
Purification Method Antigen affinity chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10068, 16068, 84388
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.