Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-18R alpha/IL-1 R5 Antibody (112614), DyLight 550, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB1216L
This item is not returnable.
View return policy
Description
IL-18R alpha/IL-1 R5 Monoclonal antibody specifically detects IL-18R alpha/IL-1 R5 in Mouse samples. It is validated for Western Blot, Flow CytometrySpecifications
IL-18R alpha/IL-1 R5 | |
Monoclonal | |
DyLight 550 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8809 | |
Store at 4°C in the dark. | |
IgG1 |
Western Blot, Flow Cytometry | |
112614 | |
50mM Sodium Borate | |
Mouse myeloma cell line NS0-derived recombinant mouse IL-18 R alpha/IL-1 R5, Ser19-Gly326, Accession # Q61098 | |
0.5 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction