Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-19 Antibody (152112), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB1035L
This item is not returnable.
View return policy
Description
IL-19 Monoclonal antibody specifically detects IL-19 in Human samples. It is validated for Flow CytometrySpecifications
IL-19 | |
Monoclonal | |
DyLight 550 | |
IL-10C, IL-19, interleukin 19, MDA1, melanoma differentiation associated protein-like protein, NG.1Melanoma differentiation-associated protein-like protein, ZMDA1interleukin-19 | |
E. coli-derived recombinant human IL-19, Leu25-Ala177, Accession # AAN40906 | |
0.5 mL | |
Biologically Active Proteins, Cytokine Research | |
29949 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry | |
152112 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction