Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-19 Monoclonal Antibody (C9)

Catalog No. PIMA543694
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA543694 20 μL
PIMA541618 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIMA543694 Supplier Invitrogen™ Supplier No. MA543694
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

IL-19 Monoclonal Antibody for Western Blot, IHC (P)

The protein encoded by this gene is a cytokine that belongs to the IL10 cytokine subfamily. This cytokine is found to be preferentially expressed in monocytes. It can bind the IL20 receptor complex and lead to the activation of the signal transducer and activator of transcription 3 (STAT3). A similar cytokine in mouse is reported to up-regulate the expression of IL6 and TNF-alpha and induce apoptosis, which suggests a role of this cytokine in inflammatory responses. Alternatively spliced transcript variants encoding the distinct isoforms have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-19
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene IL19
Gene Accession No. Q9UHD0
Gene Alias il 19; IL-10C; IL19; IL-19; ILN; Interleukin; interleukin 19; Interleukin19; interleukin-19; MDA1; MDA-1; melanoma differentiation associated protein-like protein; melanoma differentiation-associated protein-like protein; NG.1; ZMDA1
Gene Symbols IL19
Host Species Mouse
Immunogen Recombinant protein Interleukin 19. The antigen corresponds to amino acid range 25-177 of the target protein.
Purification Method Protein A/G
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29949
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.