Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1ra/IL-1F3/IL1RN Antibody (004), DyLight 594

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289280DL594
Description
IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
IL-1ra/IL-1F3/IL1RN | |
Monoclonal | |
DyLight 594 | |
DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-1ra/IL-1F3/IL1RN (NP_776214.1; Arg26-Glu177). | |
0.1 mL | |
Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
3557 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, Immunofluorescence | |
4 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction