Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1ra/IL-1F3/IL1RN Antibody (SR1317), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32168950UL
Description
IL-1ra/IL-1F3/IL1RN Monoclonal antibody specifically detects IL-1ra/IL-1F3/IL1RN in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry, SerumSpecifications
IL-1ra/IL-1F3/IL1RN | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
SR1317 | |
Western Blot 1:5000-1:20000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:50, Immunocytochemistry, Serum 1:50-1:200 | |
DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
A synthesized peptide derived from human IL-1ra/IL-1F3/IL1RN (Uniprot #: P18510) | |
50 μg | |
Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
3557 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction