Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-1ra/IL-1F3/IL1RN Mouse anti-Human, Clone: 2D11, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP311222
Description
IL-1ra/IL-1F3/IL1RN Monoclonal specifically detects IL-1ra/IL-1F3/IL1RN in Human samples. It is validated for Western Blot, Immunohistochemistry-Frozen, Immunoassay.Specifications
IL-1ra/IL-1F3/IL1RN | |
Monoclonal | |
0.1 mg/ml | |
Western Blot, Immunohistochemistry-Frozen, Immunoassay | |
DIRA, ICIL-1RA, IL1F3, IL-1ra, IL1RA, IL-1ra3, IL1RN, IL-1RN, interleukin 1 receptor antagonist, IRAP, MVCD4 | |
The antibody reacts specificly with Human IL-1ra3. | |
50 μg | |
Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Innate Immunity | |
3557 | |
Store at 4°C. Do not freeze. | |
IgG1 |
Western Blot, Immunohistochemistry (Frozen), Immunoassay | |
2D11 | |
Unconjugated | |
0.2 um filtered solution in PBS, 0.1% BSA | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction