Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IL-1RAcP Monoclonal Antibody (7E4G1E8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA529351
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human IL1R3/IL1RAP.
This gene encodes a component of the interleukin 1 receptor complex, which initiates signaling events that result in the activation of interleukin 1-responsive genes. Alternative splicing of this gene results in membrane-bound and soluble isoforms differing in their C-terminus. The ratio of soluble to membrane-bound forms increases during acute-phase induction or stress.
Specifications
IL-1RAcP | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9NPH3 | |
IL1RAP | |
Recombinant Human IL1R3 protein (Met1-Glu359). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA, Flow Cytometry | |
7E4G1E8 | |
Unconjugated | |
IL1RAP | |
6430709H04Rik; AI255955; AV239853; C3orf13; hide; IL 1R 3; IL 1R3; IL 1RAcP; IL 1RAP; IL-1 receptor accessory protein; IL1R3; IL-1R3; IL-1R-3; IL1RAcP; IL-1RAcp; Il1racpb; Il1rap; ILN; Interleukin; interleukin 1 receptor accessory protein; interleukin-1 receptor 3; Interleukin-1 receptor accessory protein; interleukin-1 receptor accessory protein beta; Interleukin-33 receptot beta chain | |
Mouse | |
Protein A | |
RUO | |
3556 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction