Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-2 Antibody (062), HRP

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290003H
Description
IL-2 Monoclonal specifically detects IL-2 in Human samples. It is validated for ELISA.Specifications
IL-2 | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
62 | |
ELISA | |
aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-2 (NP_000577.2; Ala 21-Thr153, Leu 38 Met, Leu 39 Ser, Cys 145 Ala). | |
0.1 mL | |
Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers | |
3558 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction