Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-2 Antibody (100312), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6521MFV610
Description
IL-2 Monoclonal antibody specifically detects IL-2 in Porcine samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
| IL-2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Pig | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 100312 | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
| E. coli-derived recombinant porcine IL-2, Ala21-Thr154, Accession # P26891 | |
| 0.1 mL | |
| Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers | |
| 3558 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction