Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-2 Antibody (2171A), HRP, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10356H
Description
IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOFSpecifications
IL-2 | |
Monoclonal | |
HRP | |
PBS | |
Rabbit | |
0.1 mL | |
Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers | |
3558 | |
Store at 4C in the dark. |
Flow Cytometry, Immunocytochemistry, CyTOF | |
2171A | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
E. coli-derived human IL-2, Ala21-Thr153, Accession # P60568 | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction