Learn More
Invitrogen™ IL-2 Monoclonal Antibody (8D5)

Description
MA5-17097 targets IL-2 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-17097 immunogen is purified recombinant fragment of IL2 expressed in E. Coli. MA5-17097 detects IL-2 which has a predicted molecular weight of approximately 17.6kDa.
Specifications
Specifications
| Antigen | IL-2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 8D5 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | ascites with 0.03% sodium azide |
| Gene | IL2 |
| Gene Accession No. | P60568 |
| Gene Alias | aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.