Learn More
Invitrogen™ IL-2 Monoclonal Antibody (AB12-3G4), Functional Grade, eBioscience™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ 16702738

Description
Description: This AB12-3G4 monoclonal antibody reacts with and inhibits the bioactivity of human IL-2. IL-2 is a 15 kDa cytokine expressed by B cells, T cells, and dendritic cells. IL-2 signals via a heterotrimeric receptor (CD25) and results in the activation of Jak/STAT pathways. IL-2 plays a critical role in the proliferation, activation, and differentiation of T cells. The development of CD4+CD25+ regulatory T cells is especially dependent on the presence of IL-2, making it an important immunoregulatory cytokine. Applications Reported: This AB12-3G4 monoclonal antibody inhibits the bioactivity of human IL-2. Applications Tested: The ND50 of AB12-3G4, as determined by the neutralization of Human IL-2-induced proliferation of the CTLL-2 cell line, is less than or equal to 60 ng/mL in the presence of 10 ng/mL Human IL-2 Recombinant Protein. The neutralization dose of this antibody will vary depending on cell type, assay method, and cytokine concentration. Storage and handling: Use in a sterile environment. Filtration: 0.2 μm post-manufacturing filtered. Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
IL-2 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.2 | |
P60568 | |
IL2 | |
Affinity chromatography | |
RUO | |
3558 | |
4°C | |
Liquid |
Functional Assay, Neutralization | |
AB12-3G4 | |
Functional Grade | |
IL2 | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
Mouse | |
5 mg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.