Learn More
Invitrogen™ IL-2 Monoclonal Antibody (JES6-5H4), eBioscience™
Rat Monoclonal Antibody
Supplier: Invitrogen™ BMS180

Description
Description: BMS180 (JES6-5H4) is raised against recombinant full length IL-2 of mouse origin and recommended for detection of recombinant and native IL-2 of mouse origin by IP,ELISA, FC, WB. The antibody was purified from tissue culture supernatant by protein G column chromatography. Applications Tested: ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blotting.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
IL-2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with no preservative; pH 7.4 | |
P04351 | |
IL2 | |
Rec. Mouse IL-2. | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot | |
JES6-5H4 | |
Unconjugated | |
IL2 | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
Rat | |
Affinity chromatography | |
RUO | |
16183 | |
4°C | |
Liquid |
Safety and Handling
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.