Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-2 Monoclonal Antibody (MQ1-17H12), PE, eBioscience™, Invitrogen™

Catalog No. 5011127
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
100 Tests
25 Tests
50 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-111-27 100 μg
50-111-26 50 μg
50-111-25 25 Tests
50-112-2373 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 50-111-27 Supplier Invitrogen™ Supplier No. 12702982
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Description: The MQ1-17H12 antibody reacts with human interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. The MQ1-17H12 antibody is a non-neutralizing antibody. Applications Reported: MQ1-17H12 has been reported for use in intracellular flow cytometric analysis. Applications Tested: This MQ1-17H12 antibody is offered in 2 formats: - μg size: has been tested by intracellular staining of stimulated normal human perioheral blood cells. This can be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. - test size: has been pre-titrated and tested by intracellular staining of stimulated normal human perioheral blood cells. This can be used at 5 μL (0.125 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Applications Flow Cytometry
Classification Monoclonal
Clone MQ1-17H12
Concentration 0.2 mg/mL
Conjugate PE
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene IL2
Gene Accession No. P60568
Gene Alias aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Gene Symbols IL2
Host Species Rat
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3558
Target Species Human
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.