Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-2 Mouse anti-Human, Clone: B-G5, Novus Biologicals™

Mouse Monoclonal Antibody
$423.50 - $791.00
Specifications
Antigen | IL-2 |
---|---|
Clone | B-G5 |
Concentration | 1 mg/mL |
Dilution | Western Blot, Flow Cytometry, ELISA, Functional, ELISPOT |
Applications | Western Blot, Flow Cytometry, ELISA, Functional Assay |
Description
IL-2 Monoclonal antibody specifically detects IL-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Functional Assay, ELISPOTSpecifications
IL-2 | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Functional Assay | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers | |
PBS. Sterile-filtered through 0.22 um. | |
3558 | |
IgG1 κ | |
Affinity purified | |
Antibody |
B-G5 | |
Western Blot, Flow Cytometry, ELISA, Functional, ELISPOT | |
Monoclonal | |
Purified | |
RUO | |
Human | |
aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
Natural human IL-2 | |
Primary | |
Store at 4°C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title