Learn More
Invitrogen™ IL-2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ P600
Description
P600 targets IL-2 in ELISA applications and shows reactivity with Human samples. The P600 immunogen is recombinant human IL-2. P600 detects IL-2 which has a predicted molecular weight of approximately 15 kDa. The P600 IL2 polyclonal antibody has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody M600B (biotinylated conjugate of clone BG5). Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection. Antibody P600 and biotinylated antibody M600B (clone BG5) have successfully been used in combination with recombinant IL2 protein SIL2 in ELISA applications.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
Specifications
IL-2 | |
Polyclonal | |
Unconjugated | |
IL2 | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
Rabbit | |
Protein G | |
RUO | |
3558 | |
-20°C | |
Liquid |
ELISA | |
1.0 mg/mL | |
PBS with no preservative | |
P60568 | |
IL2 | |
Recombinant human IL-2. | |
1 mg | |
Primary | |
Human | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.