Learn More
IL-2 Polyclonal Antibody, Biotin, eBioscience™, Invitrogen™
Rabbit Polyclonal Antibody
$164.00 - $495.00
Specifications
Antigen | IL-2 |
---|---|
Concentration | 0.5 mg/mL |
Applications | ELISA |
Classification | Polyclonal |
Conjugate | Biotin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-121-53
![]() |
Invitrogen™
13702881 |
50 μg |
Each for $164.00
|
|
|||||
50-112-2569
![]() |
Invitrogen™
13702885 |
500 μg |
Each for $495.00
|
|
|||||
Description
Description: The rabbit anti-human IL-2 polyclonal antibody reacts with human interleukin-2 (IL-2), a 17 kDa T cell growth factor and a major immunoregulatory cytokine. Applications Reported: This polyclonal antibody has been reported for use in detection of human IL-2 by ELISA. Applications Tested: The biotinylated polyclonal rabbit anti-human IL-2 antibody has been tested as the detection antibody in a sandwich ELISA for analysis of human IL-2 in combination with the purified MQ1-17H12 (14-7029) antibody for capture and recombinant human IL-2 (14-8029) as the standard. A suitable range of concentrations of this antibody for ELISA detection is 0.5-2 μg/mL. A standard curve consisting of doubling dilutions of the recombinant standard over the range of 1000 pg/mL - 8 pg/mL should be included in each ELISA plate. Filtration: 0.2 μm post-manufacturing filtered.
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an μgcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
IL-2 | |
ELISA | |
Biotin | |
Rabbit | |
Human | |
P60568 | |
3558 | |
IgG | |
Affinity chromatography | |
Antibody |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF | |
IL2 | |
Primary | |
4°C, do not freeze | |
IL2 |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.