Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PeproTech IL-2 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Catalog No. 500P274BT25
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 μg
50 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
500P274BT25 25 μg
500P274BT50 50 μg
50-271-8547 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 500P274BT25 Supplier PeproTech Supplier No. 500P274BT25UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

AA Sequence of recombinant protein: MAPTSSPAKE TQQHLEQLLL DLQVLLRGID NYKNLKLPMM LTFKFYLPKQ ATELKHLQCL ENELGALQRV LDLTQSKSFH LEDAGNFISN IRVTVVKLKG SENKFECQFD DEPATVVEFL RRWIAIAQSI ISTM. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Rat IL-2. Anti-Rat IL-2-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Rat IL-2 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Rat IL-2 (500-P274) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Rat IL-2. Western Blot: To detect Rat IL-2 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant Rat IL-2 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P274BT-1MG will be provided as 2 x 500 μg

Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1-1.0 mg/mL
Conjugate Biotin
Formulation PBS with no preservative
Gene IL2
Gene Accession No. P17108
Gene Alias aldesleukin; cytokine; H-IL-2; il 2; Il2; Il-2; ILN; Interleukin; interleukin 2; interleukin 2 precursor; interleukin 2 variant IL2delta2; Interleukin2; Interleukin-2; interleukin-2 precursor; interleukin-2 precursor protein; interleukin-2; interleukin 2; Involved in regulation of T cell clonal expansion; involved in regulation of T-cell clonal expansion; lymphokine; M-IL-2; patial sequence of Interleukin-2; POIL2; porcine Interleukin 2; T cell growth factor; T-cell growth factor; T-cell growth factor (TCGF); TCGF
Gene Symbols IL2
Host Species Rabbit
Immunogen E.coli-derived Recombinant Rat IL-2.
Purification Method Antigen affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116562
Target Species Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.