Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-2 R beta Antibody (TM-b1) - Azide and BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | IL-2 R beta |
---|---|
Clone | TM-b1 |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready, Neutralization |
Applications | Flow Cytometry, Immunoprecipitation, CyTOF, Neutralization |
Description
IL-2 R beta Monoclonal specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, CyTOF-ready, Block/Neutralize.Specifications
IL-2 R beta | |
0.5 mg/mL | |
Flow Cytometry, Immunoprecipitation, CyTOF, Neutralization | |
Unconjugated | |
Rat | |
Mouse | |
CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75 | |
IL2RB | |
IgG2b | |
Protein A or G purified |
TM-b1 | |
Flow Cytometry 1:10 - 1:1000, Immunoprecipitation 1:10 - 1:500, CyTOF-ready, Neutralization | |
Monoclonal | |
Purified | |
RUO | |
Aqueous with No Preservative | |
3560 | |
The immunogen for this antibody was Purified CD122. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title