Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-2 Antibody (LNKB-2), FITC, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281047F
Description
IL-2 Monoclonal specifically detects IL-2 in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| IL-2 | |
| Monoclonal | |
| FITC | |
| PBS | |
| IL2 | |
| Recombinant monoclonal antibody to IL-2. | |
| 0.1 mL | |
| Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers | |
| 3558 | |
| Human | |
| Purified |
| ELISA, Immunofluorescence | |
| LNKB-2 | |
| ELISA, Immunocytochemistry/Immunofluorescence | |
| aldesleukin, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Recognises IL-2. | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction