Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-2 Rabbit anti-Mouse, Clone: 257, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290780100UL
Description
IL-2 Monoclonal specifically detects IL-2 in Mouse samples. It is validated for ELISA.Specifications
IL-2 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
ELISA | |
257 | |
ELISA 1:5000-1:10000 | |
aldesleukin, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, T cell growth factor, T-cell growth factor, TCGF | |
Recombinant Mouse IL-2 Protein | |
100 μg | |
Adaptive Immunity, Biologically Active Proteins, Cytokine Research, Diabetes Research, Immunology, Innate Immunity, Mast Cell Markers | |
3558 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction