Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-20 Antibody (044), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290162MFV610
Description
IL-20 Monoclonal antibody specifically detects IL-20 in Human samples. It is validated for ELISASpecifications
IL-20 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
044 | |
Sandwich ELISA Capture | |
Cytokine Zcyto10, IL10D, IL-20ZCYTO10four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human IL-20 (Uniprot#: Q9NYY1; Leu25-Glu176). | |
0.1 mL | |
Biologically Active Proteins, Signal Transduction | |
50604 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction