Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-20 R beta/FNDC6 Antibody (20RNTC), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200466AF532
Description
IL-20 R beta/FNDC6 Monoclonal specifically detects IL-20 R beta/FNDC6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
IL-20 R beta/FNDC6 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
IL20RB | |
The immunogen for this antibody is FNDC6 (IL-20R2). | |
0.1 mL | |
53833.0 | |
Store at 4°C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
20RNTC | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923 | |
Rat | |
Protein A or G purified | |
Primary | |
Human, Mouse | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction