Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-20 R beta/FNDC6 Antibody (20RNTC) - BSA Free, Novus Biologicals™

Rat Monoclonal Antibody
$499.50
Specifications
Antigen | IL-20 R beta/FNDC6 |
---|---|
Clone | 20RNTC |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry 1-10 ug/ml, Immunocytochemistry/Immunofluorescence 1-10 ug/ml |
Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP20046610
![]() |
Novus Biologicals
NBP200466100UG |
100 μg |
Each of 1 for $499.50
|
|
|||||
Description
IL-20 R beta/FNDC6 Monoclonal specifically detects IL-20 R beta/FNDC6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
IL-20 R beta/FNDC6 | |
0.5 mg/mL | |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
RUO | |
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide | |
53833 | |
The immunogen for this antibody is FNDC6 (IL-20R2). | |
Primary | |
Store at 4C. Do not freeze. |
20RNTC | |
Flow Cytometry 1-10 ug/ml, Immunocytochemistry/Immunofluorescence 1-10 ug/ml | |
Monoclonal | |
Rat | |
Human, Mouse | |
FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923 | |
IL20RB | |
IgG2a κ | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title