Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-20 R beta/FNDC6 Antibody (20RNTC), DyLight 350, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP200466UV
Description
IL-20 R beta/FNDC6 Monoclonal specifically detects IL-20 R beta/FNDC6 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
IL-20 R beta/FNDC6 | |
Monoclonal | |
DyLight 350 | |
FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923 | |
Rat | |
Affinity Purified | |
RUO | |
53833 | |
Store at 4C in the dark. |
Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
20RNTC | |
Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
IL20RB | |
The immunogen for this antibody is FNDC6 (IL-20R2). | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction