Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-20 R beta/FNDC6 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298428PCP
Description
IL-20 R beta/FNDC6 Polyclonal antibody specifically detects IL-20 R beta/FNDC6 in Rat samples. It is validated for Western Blot, ELISASpecifications
IL-20 R beta/FNDC6 | |
Polyclonal | |
Western Blot, ELISA | |
FNDC6, DIRS1, IL-20R2, IL20RB interleukin 20 receptor beta, MGC34923 | |
Produced in rabbits immunized with purified, recombinant Rat IL-20 R beta/FNDC6 (Uniprot#: B8K1X9; Met1-Pro227). | |
0.1 mL | |
Primary | |
Rat | |
Purified |
Western Blot, ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
53833 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction