Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IL-20 Rabbit anti-Human, Clone: 17, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290161100UL
Description
IL-20 Monoclonal specifically detects IL-20 in Human samples. It is validated for ELISA.Specifications
IL-20 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
17 | |
ELISA 1:5000-1:10000 | |
Cytokine Zcyto10, IL10D, IL-20ZCYTO10four alpha helix cytokine, interleukin 20, interleukin-20, MGC96907 | |
Recombinant Human IL-20 Protein | |
100 μg | |
Biologically Active Proteins, Signal Transduction | |
50604 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction