Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IL-20 Recombinant Rabbit Monoclonal Antibody (044)
GREENER_CHOICE

Catalog No. PIMA530802
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530802 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530802 Supplier Invitrogen™ Supplier No. MA530802
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human interleukin-20.

Cytokines that signal through Class II receptors form a distinct family that includes the interferons and interleukin 10 (IL-10). Recent identification of several IL-10 homologs has defined a cytokine subfamily that includes AK155, IL-19, IL-20, IL-22, and IL-24. IL-20 belongs to the IL-10 family and plays a role in skin inflammation and the development of hematopoietic cells. Little is known about its other biological functions and clinical implications. These cytokines display pro-inflammatory activities in different tissues, including skin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-20
Applications ELISA
Classification Recombinant Monoclonal
Clone 044
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Il20
Gene Accession No. Q9NYY1
Gene Alias cytokine Zcyto10; four alpha helix cytokine; four alpha helix cytokine ZCYTO10; four helix bundle cytokine 10; il 20; IL10D; Il20; IL-20; ILN; Interleukin; interleukin 20; interleukin-20; MGC96907; PRO1801; UNQ852; UNQ852/PRO1801; ZCYTO10
Gene Symbols Il20
Host Species Rabbit
Immunogen Recombinant Human interleukin-20 Protein (Leu25-Glu176).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50604
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.